• USA Home
  • D195 - D4,2 Dopamine Receptor human

EMAIL THIS PAGE TO A FRIEND
D195 Sigma

D4,2 Dopamine Receptor human

membrane suspension

Purchase

Properties

recombinant   expressed in CHO cells
form   membrane suspension
packaging   vial of 50 units
storage temp.   −70°C

Description

Analysis Note

Recommended assay conditions are supplied on the product data sheet. This is a bioassayed product.

Physical form

Membrane suspension in 10 mM Tris-HCl at pH 7.2, containing 2 mM EDTA. Actual concentrations and specific binding capacity are provided with each lot

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?