• USA Home
  • D2064 - Dequalinium analog, C-14 Linker

EMAIL THIS PAGE TO A FRIEND
D2064 Sigma

Dequalinium analog, C-14 Linker

Synonym: 1,1′-(1,14-Tetradecanediyl)bis[4-amino-2-methylquinollinium] diiodide, 1,1′-(1,14-tetradecanediyl) bis diiodide, C14 Linker, DECA-14, Dequalinium analog, Dequalinium-14, Quinolinium

Purchase

Description

Biochem/physiol Actions

Protein kinase Cα (PKCα) inhibitor; antitumor agent.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?