• USA Home
  • D206 - S(−)-DS 121 hydrochloride

EMAIL THIS PAGE TO A FRIEND
D206 Sigma

S(−)-DS 121 hydrochloride

solid

Synonym: S(−)-3-(1-Propyl-3-piperidinyl)benzonitrile hydrochloride-hemicarbonate salt

  • Empirical Formula (Hill Notation) C15H20N2 · HCl · 0.5HCO3

  • Molecular Weight 295.30

  •  MDL number MFCD00671779

  •  PubChem Substance ID 24278811

Purchase

Properties

Related Categories Antagonists, Cell Biology, Cell Signaling and Neuroscience, Dopaminergics, Neuroscience,
InChI Key   AZJNIMLZMPQSPE-XFULWGLBSA-N
form   solid
optical activity   [α]22/D −10.6°, c = 1.2 in methanol(lit.)
color   white
solubility   H2O: >10 mg/mL
  methanol: >10 mg/mL
Gene Information   human ... DRD3(1814)

Description

Biochem/physiol Actions

Dopamine autoreceptor antagonist with some selectivity for the D3 dopamine receptor.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?