• USA Home
  • D2138 - Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham

EMAIL THIS PAGE TO A FRIEND
D2138 Sigma

Dulbecco’s Modified Eagle’s Medium/Nutrient Mixture F-12 Ham

Synonym: DME/F-12, 1:1 mixture

Purchase

Properties

storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?