• USA Home
  • D216 - Anti-Dopamine-β-Hydroxylase (C-Terminal) antibody produced in sheep

EMAIL THIS PAGE TO A FRIEND
D216 Sigma

Anti-Dopamine-β-Hydroxylase (C-Terminal) antibody produced in sheep

affinity isolated antibody, buffered aqueous glycerol solution

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Neurobiology, Antibodies for Neurotransmission, Antibodies for Neurotransmitter Synthesis and Metabolism,
species reactivity   human, bovine, rabbit, monkey
application(s)   immunohistochemistry: suitable
  western blot: 1:1,000
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous glycerol solution
mol wt   antigen mol wt 70-75 kDa (single or double band in samples of SDS-solubilized, DTT-reduced human adrenal medulla)
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... DBH(1621)
biological source   sheep
conjugate   unconjugated

Description

Immunogen

synthetic peptide from the C-terminal region of human dopamine-β-hydroxylase.

Physical form

Solution in 10 mM HEPES (pH 7.5), 150 mM NaCl 0.1 mg/ml BSA and 50% glycerol

Specificity

By immunoblotting, there is no cross-reactivity with adrenal medulla, sheep, dog, or rat. By immuno­­histo­chemistry, staining can be seen in the locus coeruleus of formaldehyde-fixed, cryopreserved human and monkey brain.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?