• USA Home
  • D223 - N,N-Dimethylsphingosine

EMAIL THIS PAGE TO A FRIEND
D223 Sigma

N,N-Dimethylsphingosine

oil

Synonym: (2S,3R,4E)-2-(Dimethylamino)-4-octadecene-1,3-diol, 2-Dimethylamino-4-octadecene-1,3-diol, D-erythro-N,N-dimethylsphingosine, D-Erythro-N,N-Dimethylsphingosine, DMS, Dimethylsphingosine, N,N-Dimethylsphingosine

  • CAS Number 119567-63-4

  • Empirical Formula (Hill Notation) C20H41NO2

  • Molecular Weight 327.55

  •  MDL number MFCD02259309

Purchase

Properties

InChI Key   YRXOQXUDKDCXME-YIVRLKKSSA-N
form   oil
solubility   DMSO: soluble
  ethanol: soluble
storage temp.   −20°C

Description

Physical form

Clear oil

Biochem/physiol Actions

N,N-Dimethylsphingosine (DMS) is an endogenous metabolite of sphingosine produced in various tissues and tumor cell lines. N,N-Dimethylsphingosine is a potent inhbitor of sphingosine kinase. DMS levels are elevated in the spinal cord, and contributes to allodynia, in a transected-nerve model of neuropathic pain.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 2
WGK Germany 
1
Flash Point(F) 
57 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?