• USA Home
  • D2250 - Nα-Dansyl-L-tryptophan cyclohexylammonium salt

EMAIL THIS PAGE TO A FRIEND
D2250 Sigma

Nα-Dansyl-L-tryptophan cyclohexylammonium salt

  • CAS Number 19461-29-1

  • Empirical Formula (Hill Notation) C23H23N3O4S · C6H13N

  • Molecular Weight 536.69

  •  MDL number MFCD00069706

  •  PubChem Substance ID 24893668

Purchase

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?