• USA Home
  • D2255 - O-Mono-2,4-DNP-L-tyrosine

EMAIL THIS PAGE TO A FRIEND
D2255 Sigma

O-Mono-2,4-DNP-L-tyrosine

  • CAS Number 10567-73-4

  • Empirical Formula (Hill Notation) C15H13N3O7

  • Molecular Weight 347.28

  •  MDL number MFCD00038108

  •  PubChem Substance ID 24893669

Purchase

Properties

Related Categories Amino Acid Analysis, Amino Acid Analysis Reagents, Biochemicals and Reagents, Peptide Analysis and Characterization, Peptides More...
InChI Key   OHFDOVYRFJQGIR-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?