• USA Home
  • D2439 - D4.2 Dopamine Receptor human

EMAIL THIS PAGE TO A FRIEND
D2439 Sigma

D4.2 Dopamine Receptor human

recombinant, expressed in baculovirus infected Sf9 cells, membrane suspension

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuroscience, Neurotransmission, Neurotransmitter Receptors More...
recombinant   expressed in baculovirus infected Sf9 cells
form   membrane suspension
packaging   vial of 50 units (250 μl)
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... DRD2(1813), DRD4(1815)

Description

Analysis Note

Recommended assay conditions are supplied on the product data sheet.

Caution

Membranes retain original specific activity for several months when stored in original packaging solution.

Packaging

Actual concentration and specific binding capacity are provided with each lot.

Unit Definition

One unit will yield specific binding of ~1,000 dpm at a concentration of radioligand comparable to that of the Kd.

Physical form

Membrane suspension in 50 mM Tris-HCl, pH 7.4, 10% glycerol, and 1% bovine serum albumin.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?