• USA Home
  • D2519 - 4,5-Dichloro-o-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
D2519 Sigma

4,5-Dichloro-o-phenylenediamine

Synonym: 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichloro-1,2-phenylenediamine

Purchase

Properties

InChI Key   IWFHBRFJOHTIPU-UHFFFAOYSA-N
mp   158-164 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D71607 4,5-Dichloro-o-phenylenediamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?