• USA Home
  • D2630 - 2,4-Dinitrophenylhydrazine

EMAIL THIS PAGE TO A FRIEND
D2630 Sigma

2,4-Dinitrophenylhydrazine

  • CAS Number 119-26-6

  • Linear Formula (O2N)2C6H3NHNH2

  • Molecular Weight 198.14

  •  Beilstein Registry Number 615586

  •  MDL number MFCD00007578

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   HORQAOAYAYGIBM-UHFFFAOYSA-N
contains   ≥30% water as stabilizer (to comply with DOT regulations)
mp   197-200 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3380 - class 4.1 - PG 1 - DOT UN1325Desensitized explosive, solid, n.o.s. (2,4-DINITROPHENYLHYDRAZINE, WETTED), HI: all
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D199303 2,4-Dinitrophenylhydrazine, reagent grade, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?