• USA Home
  • D2681 - 2-Deoxy-2-phthalimido-3,4,6-tri-O-acetyl-α-D-galactopyranosyl fluoride

EMAIL THIS PAGE TO A FRIEND
D2681 Sigma

2-Deoxy-2-phthalimido-3,4,6-tri-O-acetyl-α-D-galactopyranosyl fluoride

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates D-F, Monosaccharide More...
InChI Key   YRVBOYANNCZRHU-SFFUCWETSA-N
impurities   ≤10% β-anomer

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?