• USA Home
  • D2770 - (S)-(−)-N,N-Dimethyl-1-phenethylamine

EMAIL THIS PAGE TO A FRIEND
D2770 Sigma

(S)-(−)-N,N-Dimethyl-1-phenethylamine

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
2
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?