• USA Home
  • D2900 - Dolichos biflorus (Horse gram) acetone powder

EMAIL THIS PAGE TO A FRIEND
D2900 Sigma

Dolichos biflorus (Horse gram) acetone powder

Purchase

Description

Other Notes

Contains anti-A1 lectin

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?