• USA Home
  • D2907 - 1,2-Didodecanoyl-3-tetradecanoyl-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
D2907 Sigma

1,2-Didodecanoyl-3-tetradecanoyl-rac-glycerol

≥99%

Synonym: 1,2-Dilauroyl-3-myristoyl-rac-glycerol

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerolipids, Glycerophospholipid Metabolism, Lipids,
InChI Key   SMUUMIRCIBXLGI-UHFFFAOYSA-N
assay   ≥99%
storage temp.   −20°C

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?