• USA Home
  • D3150 - Deoxyribonuclease, ATP dependent from Micrococcus luteus (lysodeikticus)

EMAIL THIS PAGE TO A FRIEND
D3150 Sigma

Deoxyribonuclease, ATP dependent from Micrococcus luteus (lysodeikticus)

2,000-10,000 units/mL, buffered aqueous glycerol solution

Synonym: Exodeoxyribonuclease V, Exonuclease V

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
form   buffered aqueous glycerol solution
concentration   2,000-10,000 units/mL
shipped in   wet ice
storage temp.   −20°C

Description

Unit Definition

One unit will produce 10 nanomoles of acid soluble nucleotides from 20 nanomoles of e. coli DNA in 30 min at pH 9.4 at 37°C.

Physical form

Solution in 50% glycerol containing 20 mM Tris HCl, pH 7.5, 10 mM 2-mercaptoethanol, 0.1 mM EDTA and 500 μg per mL bovine serum albumin

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?