• USA Home
  • D3259 - Deoxyribonucleic acid-methyl green standardization set

EMAIL THIS PAGE TO A FRIEND
D3259 Sigma

Deoxyribonucleic acid-methyl green standardization set

Purchase

Properties

storage temp.   −20°C

Description

Application

For standardizing DNA

Price and Availability

Standard component

Description

Product #

Add to Cart

DNA-Methyl Green 100 mg SDS    
DNase I 5 vials SDS    
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

D2376 Deoxyribonucleic acid methyl green, DNAse substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?