| Related Categories | Cryoware, Glass Dewars, Labware, Wide-mouth Dewars with Silvered Glass Liner |
| capacity | 2 L |
| inside dimensions D × H | 4.2 in. (106 mm) × 8.1 in. (205 mm) |
| outside dimensions D × H | 5.6 in. (142 mm) × 10.5 in. (268 mm) |
Straight sides, a swing-over carrying handle, and no neck constriction; suitable for benchtop immersion applications. Vented insulating lids clamp to the body of the flask.
Please note: The manufacturer suggests rinsing the unit each day with distilled water to prevent the seals from deteriorating.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?