• USA Home
  • D3388 - DL-Decanoylcarnitine chloride

EMAIL THIS PAGE TO A FRIEND
D3388 Sigma

DL-Decanoylcarnitine chloride

  • CAS Number 18822-87-2

  • Empirical Formula (Hill Notation) C17H34ClNO4

  • Molecular Weight 351.91

  •  MDL number MFCD00070304

  •  PubChem Substance ID 24893749

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   MREFADZXZKRPGZ-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?