EMAIL THIS PAGE TO A FRIEND
D3408 Sigma

Dansyl-D-Ala-Gly

~95%

  • CAS Number 92175-75-2

  • Empirical Formula (Hill Notation) C17H21N3O5S

  • Molecular Weight 379.43

  •  MDL number MFCD00214269

  •  PubChem Substance ID 24893751

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   IKCPKYTWSUFCRD-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?