• USA Home
  • D3547 - β-Waste disposal container, Nalgene®
EMAIL THIS PAGE TO A FRIEND
D3547 Sigma
β-Waste disposal container, Nalgene®
inside L × W × H 16 in. (406 mm) × 16 in. (406 mm) × 29 in. (736 mm)
Purchase
Properties
Related Categories Disposal Containers, Labware, Manufacturer Browser, Nalgene, Nalgene Safety Equipment,
material   acrylic
mfr. no.   Nalgene
inside L × W × H   16 in. (406 mm) × 16 in. (406 mm) × 29 in. (736 mm)
Description
General description
Constructed of 3/8 in. clear acrylic with a large hinged door on top. Hinged corners of body allow removal of waste. Designed to hold radiation waste cartons, 20 L Nalgene carboys, or other suitable containers.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?