• USA Home
  • D3563 - Anti-DR3, C-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
D3563 Sigma

Anti-DR3, C-Terminal antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-Apo-3, Anti-Death Receptor 3, Anti-LARD, Anti-TRAMP, Anti-Wsl-1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Apoptosis, Antibodies for Cell Biology, Antibodies to Death Receptors,
species reactivity   human
application(s)   microarray: suitable
  western blot: 1 μg/mL using human Jurkat cell extract
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 59 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... TNFRSF25(8718)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 398-417 of the C-terminal of human DR3 precursor.

General description

DR3 is a member of the TNF superfamily. It is expressed in lymphocyte rich tissues such as blood, thymus and spleen.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Application

Anti-DR3 antibody produced in rabbit is suitable for microarray and immunoblotting at a working concentration of 1μg/ml using total cell lysates from Jurkat cells.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

DR3 is a death domain-containing receptor that is involved in inducing apoptosis or programmed cell death in cells. Binding of ligand to these receptors sends signals that activate members of the caspase family of proteases, which cause the degradation of chromosomal DNA by activating DNase. It also activates NF-κB, a protein complex that controls transcription of DNA.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?