EMAIL THIS PAGE TO A FRIEND
D3565 Sigma

Declere solution

20× concentrate

Purchase

Description

Reconstitution

Working solution is prepared by diluting the concentrate 1:20 with water.

Application

Single reagent treatment which can be used to deparaffinize, rehydrate, and unmask formalin-fixed, paraffin embedded tissue sections for immunohistochemical staining.

The solution was used for dewaxing/unmasking of formalin-fixed, paraffin embedded tumor tissues collected from vehicle- or chemo-treated mice at regression and relapse for immunofluorescence analysis.

Other Notes

The solution is completely biodegradable and non-toxic.

Price and Availability


Explore with Confidence

Tissue Diagnostics

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?