• USA Home
  • D3662 - Dimethylamine-boranecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
D3662 Sigma

Dimethylamine-boranecarboxylic acid

Synonym: Borodimethylglycine, Dimethylamine-carboxyborane

  • CAS Number 77356-05-9

  • Empirical Formula (Hill Notation) C3H10BNO2

  • Molecular Weight 102.93

  •  MDL number MFCD00133268

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   BTSWWBIRVRVZNU-UHFFFAOYSA-N
storage temp.   2-8°C

Description

Biochem/physiol Actions

Hypolipidemic and antineoplastic agent.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ED1170000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?