EMAIL THIS PAGE TO A FRIEND
D3764 Sigma

DEAE-Cellulose

preswollen, microgranular

Synonym: Cellulose DEAE, Diethylaminoethyl-cellulose

Purchase

Properties

InChI Key   GUBGYTABKSRVRQ-WFVLMXAXSA-N
form   preswollen, microgranular
contains   ~70% moisture content
capacity   ~0.3 meq/g(as supplied)
  ~1.0 meq/g(dry)

Description

Application

DEAE-Cellulose is used in ion exchange chromatography, protein chromatography and anion exchange media. DEAE cellulose has been used to study acute myocardial infarction

Price and Availability


Amicon Pro Purification System
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?