• USA Home
  • D3787 - 4,4-Dimethyl-2-imidazoline

EMAIL THIS PAGE TO A FRIEND
D3787 Sigma

4,4-Dimethyl-2-imidazoline

~97%

Purchase

Properties

assay   ~97%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?