• USA Home
  • D3788 - (8S,15S)-Dihydroxy-(5Z,9E,11Z,13E)-eicosatetraenoic acid

EMAIL THIS PAGE TO A FRIEND
D3788 Sigma

(8S,15S)-Dihydroxy-(5Z,9E,11Z,13E)-eicosatetraenoic acid

≥98%, ~75 μg/mL in ethanol

Synonym: (8S,15S)-DiHETE

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acids and conjugates, Fatty Acyls, Hydroxy fatty acids, Lipids,
InChI Key   NNPWRKSGORGTIM-HCCKYKKOSA-N
assay   ≥98%
concentration   ~75 μg/mL in ethanol
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Packaged under Argon.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 2
WGK Germany 
1
Flash Point(F) 
57 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?