• USA Home
  • D3913 - (±)-Deoxyephedrine-d5 hydrochloride

EMAIL THIS PAGE TO A FRIEND
D3913 Sigma

(±)-Deoxyephedrine-d5 hydrochloride

Synonym: DL-Methamphetamine-d5

Purchase

Properties

InChI Key   TWXDDNPPQUTEOV-QVBWKROHSA-N
isotopic purity   98 atom % D
form   neat
drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
mass shift   M+5

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?