EMAIL THIS PAGE TO A FRIEND
D4280 Sigma

Deuterium oxide

Synonym: Heavy water, Water-d2

Purchase

Properties

InChI Key   XLYOFNOQVPJJNP-ZSJDYOACSA-N
isotopic purity   99.9 atom % D
refractive index   n20/D 1.328(lit.)
bp   101.4 °C(lit.)
mp   3.8 °C(lit.)
density   1.107 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
ZC0230000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?