• USA Home
  • D4407 - Developer II concentrate

EMAIL THIS PAGE TO A FRIEND
D4407 Sigma

Developer II concentrate

Purchase

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1198 - class 3 - PG 3 - Formaldehyde, solutions, flammable
WGK Germany 
2
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

PROTSIL1 ProteoSilver Silver Stain Kit

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?