• USA Home
  • D4565 - 2,3-Di-nor-8-isoprostaglandin F

EMAIL THIS PAGE TO A FRIEND
D4565 Sigma

2,3-Di-nor-8-isoprostaglandin F

≥98%, methyl acetate solution

  • CAS Number 221664-05-7

  • Empirical Formula (Hill Notation) C18H30O5

  • Molecular Weight 326.43

  •  MDL number MFCD03411993

Purchase

Properties

Related Categories Arachidonic Acid Cascade, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   IDKLJIUIJUVJNR-JSEKUSAISA-N
assay   ≥98%
form   methyl acetate solution
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Metabolite of 8-iso-PGF

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1231 - class 3 - PG 2 - Methyl acetate, solution
WGK Germany 
2
Flash Point(F) 
3.2 °F
Flash Point(C) 
-16 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?