• USA Home
  • D4658 - Pulse Marker 225-2,200 kb

EMAIL THIS PAGE TO A FRIEND
D4658 Sigma

Pulse Marker 225-2,200 kb

for pulse-field DNA electrophoresis

Purchase

Properties

grade   for molecular biology
form   liquid
usage   100 uses
suitability   suitable for electrophoresis (pulsed-field DNA)
shipped in   dry ice
storage temp.   2-8°C

Description

Physical form

Solution in 0.25 M EDTA, pH 9.0, 50% glycerol

Application

The 225-2200 kb marker consists of chromosomes of Saccharamyces cerevisiae. The recommended agarose gel concentration is 1.0%. The recommended plug size is approximately 20 μl (1 μg) or two small graduations on the syringe.

Legal Information

Pulse Marker is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?