• USA Home
  • D5005 - trans-p,p′-Diphenylstilbene

EMAIL THIS PAGE TO A FRIEND
D5005 Sigma

trans-p,p′-Diphenylstilbene

suitable for liquid scintillation spectrometry

Synonym: 1,2-bis(p-Biphenylyl)ethylene, DPS

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Fluorescence/Luminescence Spectroscopy, Luminescent Compounds/Detection,
InChI Key   HXWQJYVUJPBQEW-VAWYXSNFSA-N
suitability   suitable for liquid scintillation spectrometry
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?