• USA Home
  • D5125 - 16-Dehydropregnenolone oxime

EMAIL THIS PAGE TO A FRIEND
D5125 Sigma

16-Dehydropregnenolone oxime

Synonym: 3β-Hydroxy-5,16-pregnadien-20-one oxime, 5,16-Pregnadien-3β-ol-20-one oxime

  • CAS Number 1045-71-2

  • Empirical Formula (Hill Notation) C21H31NO2

  • Molecular Weight 329.48

  •  MDL number MFCD00046221

  •  PubChem Substance ID 24893922

Purchase

Properties

InChI Key   PUAYLIXQYVQBPE-MZFALXGWSA-N
Gene Information   human ... CYP17A1(1586)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?