• USA Home
  • D5164 - 5,7-Dichloro-8-hydroxy-2-methylquinoline

EMAIL THIS PAGE TO A FRIEND
D5164 Sigma

5,7-Dichloro-8-hydroxy-2-methylquinoline

Synonym: 5,7-Dichloro-2-methyl-8-quinolinol, 5,7-Dichloro-8-hydroxyquinaldine, 5,7-Dichloro-8-quinaldinol, BCM

  • CAS Number 72-80-0

  • Empirical Formula (Hill Notation) C10H7Cl2NO

  • Molecular Weight 228.07

  •  Beilstein Registry Number 156683

  •  EC Number 200-789-3

  •  MDL number MFCD00023984

  •  PubChem Substance ID 24893930

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics A-F, Biochemicals and Reagents, Building Blocks,
InChI Key   GPTXWRGISTZRIO-UHFFFAOYSA-N
mp   108-112 °C (dec.)(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
VC5600000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?