EMAIL THIS PAGE TO A FRIEND
D5185 Sigma

Dermato supplement

for microbiology

Purchase

Properties

Related Categories Media, Microbiology, Supplements More...
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
shipped in   wet ice
storage temp.   2-8°C

Description

Components

Composition: (per vial)
Cycloheximide: 250mg
Chlortetracycline: 50mg
Gentamicin: 50mg

Application

An antibiotic supplement recommended for the selective isolation of pathogenic dermatophytes.

Reconstitution

Sufficient for 500 ml medium

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 1
WGK Germany 
3
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?