• USA Home
  • D5189 - L-Asp-Asp-Rhodamine trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
D5189 Sigma

L-Asp-Asp-Rhodamine trifluoroacetate salt

>98%, solid

Synonym: (L-Asp)2 Rhodamine 110 ⋅ TFA

  • Empirical Formula (Hill Notation) C28H24N4O9 · C2HF3O2

  • Molecular Weight 674.53

  •  MDL number MFCD03452916

Purchase

Description

Amino Acid Sequence

Asp-Asp-Rh

Application

Substrate for detection of intracellular caspase activation by flow cytometry after apoptosis induction in intact hematopoietic cell lines.

Price and Availability


Amplified Detection

All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?