EMAIL THIS PAGE TO A FRIEND
D5262 Sigma

Didansylcadaverine

Synonym: Pentane-1,5-bis(dimethylaminonaphthalenesulfonamide)

  • CAS Number 55521-24-9

  • Empirical Formula (Hill Notation) C29H36N4O4S2

  • Molecular Weight 568.75

  •  MDL number MFCD00056585

  •  PubChem Substance ID 24893932

Purchase

Properties

Related Categories Activity, Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Fluorogenic More...
InChI Key   USJKPOZTSFSPTN-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?