• USA Home
  • D5267 - 2,7-Dimethoxynaphthalene

EMAIL THIS PAGE TO A FRIEND
D5267 Sigma

2,7-Dimethoxynaphthalene

Purchase

Properties

storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?