• USA Home
  • D5280 - Dulbecco’s Modified Eagle’s Medium - low glucose

EMAIL THIS PAGE TO A FRIEND
D5280 Sigma

Dulbecco’s Modified Eagle’s Medium - low glucose

powder, Auto-Mod, cell culture tested

Synonym: DME, DMEM

Purchase

Properties

Quality Level   GMP
form   powder
IVD   for in vitro diagnostic use
suitability   cell culture tested
storage temp.   2-8°C

Description

Other Notes

Modified for Autoclaving.
With 1000 mg/L glucose. Without L-glutamine and sodium bicarbonate.

Quantity

Formulated to contain 9.6 grams of powder per liter of medium.

Reconstitution

Supplement with 0.584 g/L L-glutamine, 3.7 g/L sodium bicarbonate.

Legal Information

Auto-Mod is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation
Protocols & Articles

Related Content

Dulbecco's Modified Eagle's Medium - DME, DMEM

Many modifications of Eagle's Medium have been developed since the original formulation appeared in the literature. Among the most widely used of these modifications is Dulbecco's Modified Eagle's me...
Keywords: Cell culture, Substitutions, Vitamins

Related Products

replaced by

Product #

Description

Add to Cart

D5523 Dulbecco’s Modified Eagle’s Medium - low glucose, With 1000 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?