• USA Home
  • D5295 - 3,6-Dithiaoctane-1,8-diol

EMAIL THIS PAGE TO A FRIEND
D5295 Sigma

3,6-Dithiaoctane-1,8-diol

Synonym: 2,2′-(Ethylenedithio)diethanol, Lindlar Catalyst Poison

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   PDHFSBXFZGYBIP-UHFFFAOYSA-N
suitability   suitable for photographic applications

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?