• USA Home
  • D5378 - 1,5-Difluoro-2,4-dinitrobenzene

EMAIL THIS PAGE TO A FRIEND
D5378 Sigma

1,5-Difluoro-2,4-dinitrobenzene

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
CZ5663200

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?