• USA Home
  • D5420 - 3,5-Dinitrobenzoic acid

EMAIL THIS PAGE TO A FRIEND
D5420 Sigma

3,5-Dinitrobenzoic acid

suitable for photographic applications

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   VYWYYJYRVSBHJQ-UHFFFAOYSA-N
mp   204-206 °C(lit.)
suitability   suitable for photographic applications

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DG9140700
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?