• USA Home
  • D5793 - Deoxyribonuclease I, rnase-free

EMAIL THIS PAGE TO A FRIEND
D5793 Sigma

Deoxyribonuclease I, rnase-free

Synonym: DNase I

Purchase

Properties

shipped in   dry ice
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RF0750000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?