• USA Home
  • D6013 - 5,5-Diallylbarbituric acid

EMAIL THIS PAGE TO A FRIEND
D6013 Sigma

5,5-Diallylbarbituric acid

Synonym: Allobarbital

  • CAS Number 52-43-7

  • Empirical Formula (Hill Notation) C10H12N2O3

  • Molecular Weight 208.21

  •  MDL number MFCD00056090

  •  PubChem Substance ID 329798819

Purchase

Properties

InChI Key   FDQGNLOWMMVRQL-UHFFFAOYSA-N
form   neat
drug control   USDEA Schedule III; Home Office Schedule 3; psychotrope; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?