• USA Home
  • D6039 - Desmethyldiazepam-d5 solution

EMAIL THIS PAGE TO A FRIEND
D6039 Sigma

Desmethyldiazepam-d5 solution

100 μg/L in methanol

Synonym: 7-Chloro-1,3-dihydro-5-(phenyl-d5)-2H-1,4-benzodiazepin-2-one solution, Nordiazepam-d5 solution

Purchase

Properties

InChI Key   AKPLHCDWDRPJGD-RALIUCGRSA-N
drug control   USDEA Schedule III; Home Office Schedule 4.1; psychotrope; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   100 μg/L in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
mass shift   M+5
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?