• USA Home
  • D6168 - Monoclonal Anti-Dynein (Intermediate Chain) antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
D6168 Sigma

Monoclonal Anti-Dynein (Intermediate Chain) antibody produced in mouse

clone 1869A, ascites fluid

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, DQ-DZ, Primary Antibodies
biological source   mouse
antibody form   ascites fluid
clone   1869A, monoclonal
contains   15 mM sodium azide
species reactivity   Chlamydomonas, trout (Salmo gairdneri), sea urchin (Tripneustes gratilla), Strongylocentrotus purpuratus
application(s)   western blot: 1:10,000 using Chlamydomonas flagella
  western blot: 1:10000 using Chlamydomonas reinhardtii flagella preparation
isotype   IgG1
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Immunogen

outer arm dynein intermediate chain of Chlamydomonas.

Specificity

Recognizes the intermediate chain (IC69, 69 kDa) of Chlamydomonas dynein, but does not recognize any other protein in the axoneme. Reacts strongly with the 76 kDa intermediate chain outer arm dynein of sea urchin (Tripneustes gratilla, Strongylocentrotus purpuratus) and with a single component (IC2, 73 kDa) of trout (Salmo gairdneri) outer arm dynein. Does not recognize any component in axonemes prepared from scallop gill cilia or sheep sperm, purified 14S and 22S dyneins from cilia of Tetrahymena thermophila, or chick brain extract.

Application

May be used for domain mapping within IC69, electron microscope studies on dynein particles, and for protein-protein association studies on purified outer arm dynein and axonemes from Chlamydomonas flagella.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?