• USA Home
  • D6178 - 4-(5,6-Dimethoxy-N-phthalimidinyl)benzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
D6178 Sigma

4-(5,6-Dimethoxy-N-phthalimidinyl)benzenesulfonyl chloride

Synonym: (1,3-Dihydro-5,6-dimethoxy-1-oxo-2H-isoindol-2-yl)-4-benzenesulfonyl chloride, DPS-Cl

Purchase

Properties

Related Categories Biochemicals and Reagents, Derivatization agents, Luminescent Compounds/Detection More...
InChI Key   JGILUZFVCJOPOG-UHFFFAOYSA-N
storage temp.   −20°C

Description

Application

Fluorescent derivatizing reagent for amino acid analysis.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1759 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?