• USA Home
  • D6189 - Dendroaspis Natriuretic Peptide trifluoroacetate salt

EMAIL THIS PAGE TO A FRIEND
D6189 Sigma

Dendroaspis Natriuretic Peptide trifluoroacetate salt

≥90% (HPLC)

  • Empirical Formula (Hill Notation) C180H282N56O56S2

  • Molecular Weight 4190.64

  •  MDL number MFCD02687290

Purchase

Description

Amino Acid Sequence

Glu-Val-Lys-Tyr-Asp-Pro-Cys-Phe-Gly-His-Lys-Ile-Asp-Arg-Ile-Asn-His-Val-Ser-Asn-Leu-Gly-Cys-Pro-Ser-Leu-Arg-Asp-Pro-Arg-Pro-Asn-Ala-Pro-Ser-Thr-Ser-Ala [Disulfide Bridge: 7-23]

Biochem/physiol Actions

Structural homolog of natiuretic peptides containing a 17 amino acid disulfide ring similar to ANP, BNP, and CNP that mediate their physiological effects through particulate guanylyl cyclase receptors generating cGMP.

Price and Availability


Better Together

Discover your Target

Making Headway
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?