• USA Home
  • D6261 - β,β-Dimethylacrylic acid

EMAIL THIS PAGE TO A FRIEND
D6261 Sigma

β,β-Dimethylacrylic acid

≥99%

Synonym: 3-Methyl-2-butenoic acid, Senecioic acid

Purchase

Properties

assay   ≥99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
GQ5425000
Flash Point(F) 
199.4 °F
Flash Point(C) 
93 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?